missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. F02165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02165G 5 g
F021625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02165G Supplier TCI America Supplier No. F02165G

Chemical Identifiers

CAS 331-25-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004331
InChI Key YEAUYVGUXSZCFI-UHFFFAOYSA-N
Synonym 3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid
PubChem CID 67617
IUPAC Name 2-(3-fluorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(F)=C1

Specifications

Melting Point 47°C
Color White
Boiling Point 125°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorophenylacetic Acid