missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetic acid, 98%

CAS: 331-25-9 | C8H7FO2 | 154.14 g/mol

Supplier:  Thermo Scientific Chemicals A1563514

 View more versions of this product

Catalog No. AAA1563514


Only null left
Explore available promotions

Description

Description

3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

Solubility
Slightly soluble in chloroform and methanol. Insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

3-Fluorophenylacetic acid
44°C to 48°C
>110°C (230°F)
C8H7FO2
25 g
3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid
YEAUYVGUXSZCFI-UHFFFAOYSA-N
2-(3-fluorophenyl)acetic acid
67617
98%
331-25-9
151°C (0.8 mmHg)
Odorless
MFCD00004331
775898
Slightly soluble in chloroform and methanol. Insoluble in water.
OC(=O)CC1=CC=CC(F)=C1
154.14
154.14

Chemical Identifiers

331-25-9
154.14
YEAUYVGUXSZCFI-UHFFFAOYSA-N
67617
OC(=O)CC1=CC=CC(F)=C1
C8H7FO2
MFCD00004331
3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid
2-(3-fluorophenyl)acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-360-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only