missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetic acid, 98%

Catalog No. AAA1563522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1563522 100 g
AAA1563514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1563522 Supplier Thermo Scientific Chemicals Supplier No. A1563522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 331-25-9 | C8H7FO2 | 154.14 g/mol

3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

Solubility
Slightly soluble in chloroform and methanol. Insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 331-25-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004331
InChI Key YEAUYVGUXSZCFI-UHFFFAOYSA-N
Synonym 3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid
PubChem CID 67617
IUPAC Name 2-(3-fluorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(F)=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 151°C (0.8 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Quantity 100 g
Beilstein 775898
Solubility Information Slightly soluble in chloroform and methanol. Insoluble in water.
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 3-Fluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-360-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only