missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl isothiocyanate, 97+%

CAS: 404-72-8 | C7H4FNS | 153.17 g/mol

$105.40 - $333.64

Chemical Identifiers

CAS 404-72-8
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.17
MDL Number MFCD00004804
InChI Key HDBACITVPQEAGG-UHFFFAOYSA-N
Synonym 3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
PubChem CID 9834
IUPAC Name 1-fluoro-3-isothiocyanatobenzene
SMILES FC1=CC=CC(=C1)N=C=S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2500106
View Documents
Thermo Scientific Chemicals
B2500106
5 g
Each for $105.40
Only null left
 
AAB2500114
View Documents
Thermo Scientific Chemicals
B2500114
25 g
Each for $333.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

404-72-8
153.17
HDBACITVPQEAGG-UHFFFAOYSA-N
9834
FC1=CC=CC(=C1)N=C=S
C7H4FNS
MFCD00004804
3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
1-fluoro-3-isothiocyanatobenzene

Specifications

404-72-8
227°C
Pungent
1.62
5 g
2081172
3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
FC1=CC=CC(=C1)N=C=S
153.17
153.18
3-Fluorophenyl isothiocyanate
1.27
85°C (185°F)
C7H4FNS
MFCD00004804
UN2922
Moisture sensitive
HDBACITVPQEAGG-UHFFFAOYSA-N
1-fluoro-3-isothiocyanatobenzene
9834
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 206-967-7

RTECSNumber : NX8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only