missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl isothiocyanate, 97+%

Catalog No. AAB2500106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2500106 5 g
AAB2500114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2500106 Supplier Thermo Scientific Chemicals Supplier No. B2500106

CAS: 404-72-8 | C7H4FNS | 153.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 404-72-8
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.17
MDL Number MFCD00004804
InChI Key HDBACITVPQEAGG-UHFFFAOYSA-N
Synonym 3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
PubChem CID 9834
IUPAC Name 1-fluoro-3-isothiocyanatobenzene
SMILES FC1=CC=CC(=C1)N=C=S

Specifications

Density 1.27
Boiling Point 227°C
Flash Point 85°C (185°F)
Odor Pungent
Refractive Index 1.62
Quantity 5 g
UN Number UN2922
Beilstein 2081172
Sensitivity Moisture sensitive
Formula Weight 153.18
Percent Purity ≥97%
Chemical Name or Material 3-Fluorophenyl isothiocyanate
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 206-967-7
RTECSNumber NX8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only