missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenol, 98%

Catalog No. p-7021316
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359809 10 g
AAA1359818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1359809 Supplier Thermo Scientific Chemicals Supplier No. A1359809
Only null left

CAS: 372-20-3 | C6H5FO | 112.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-20-3
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.103
MDL Number MFCD00002254
InChI Key SJTBRFHBXDZMPS-UHFFFAOYSA-N
Synonym m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
PubChem CID 9743
IUPAC Name 3-fluorophenol
SMILES C1=CC(=CC(=C1)F)O

Specifications

Melting Point 12°C to 14°C
Density 1.246
Boiling Point 177°C to 179°C
Flash Point 71°C (159°F)
Odor Phenol-like
Refractive Index 1.514
Quantity 10 g
UN Number UN2922
Beilstein 1904536
Formula Weight 112.1
Percent Purity 98%
Chemical Name or Material 3-Fluorophenol
Show More Show Less
Hazard Category H227-H301-H312-H315-H318-H332-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 206-748-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only