missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorocatechol 98.0+%, TCI America™

Catalog No. F02821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02821G 1 g
F02825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02821G Supplier TCI America Supplier No. F02821G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 363-52-0
Molecular Formula C6H5FO2
Molecular Weight (g/mol) 128.10
MDL Number MFCD00042582
InChI Key DXOSJQLIRGXWCF-UHFFFAOYSA-N
Synonym 3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
PubChem CID 67764
ChEBI CHEBI:39876
IUPAC Name 3-fluorobenzene-1,2-diol
SMILES OC1=CC=CC(F)=C1O

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 128.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorocatechol