missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzylamine 98.0+%, TCI America™

Catalog No. p-7129740
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05395G 5 g
F053925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05395G Supplier TCI America Supplier No. F05395G

Chemical Identifiers

CAS 100-82-3
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
InChI Key QVSVMNXRLWSNGS-UHFFFAOYSA-N
Synonym 3-fluorobenzylamine, 3-fluorophenyl methanamine, m-fluorobenzylamine, benzenemethanamine, 3-fluoro, 1-3-fluorophenyl methanamine, 3-fluoro-benzylamine, benzylamine, m-fluoro, 3-fluorophenyl methylamine, m-fluorobenzyl amine, 3-fluorobenzyl amine
PubChem CID 66853
IUPAC Name (3-fluorophenyl)methanamine
SMILES C1=CC(=CC(=C1)F)CN

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 2735
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzylamine