missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzylamine, 97%

Catalog No. AC155500050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155500050 5 g Glass bottle
AC155500010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155500050 Supplier Thermo Scientific Chemicals Supplier No. 155500050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-82-3 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-82-3
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00008113
InChI Key QVSVMNXRLWSNGS-UHFFFAOYSA-N
Synonym 3-fluorobenzylamine, 3-fluorophenyl methanamine, m-fluorobenzylamine, benzenemethanamine, 3-fluoro, 1-3-fluorophenyl methanamine, 3-fluoro-benzylamine, benzylamine, m-fluoro, 3-fluorophenyl methylamine, m-fluorobenzyl amine, 3-fluorobenzyl amine
PubChem CID 66853
IUPAC Name (3-fluorophenyl)methanamine
SMILES C1=CC(=CC(=C1)F)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 87.0°C to 89.0°C (25.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5152
Linear Formula FC6H4CH2NH2
Quantity 5 g
Specific Gravity 1.09
Formula Weight 125.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 202-891-3
Recommended Storage May darken during storage

RUO – Research Use Only