missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzyl Chloride 95.0+%, TCI America™
Supplier: TCI America F021025G
Specifications
| 3-Fluorobenzyl Chloride | |
| Yellow | |
| C7H6ClF | |
| 25 g | |
| 3-fluorobenzyl chloride, 1-chloromethyl-3-fluorobenzene, m-fluorobenzyl chloride, benzene, 1-chloromethyl-3-fluoro, 3-fluorobenzylchloride, alpha-chloro-3-fluorotoluene, alpha-chloro-m-fluorotoluene, fluorobenzyl-3 chloride, toluene, .alpha.-chloro-m-fluoro, .alpha.-chloro-m-fluorotoluene | |
| C1=CC(=CC(=C1)F)CCl | |
| 144.573 | |
| 144.57 | |
| Liquid |
| 456-42-8 | |
| 180°C | |
| MFCD00000904 | |
| 2920 | |
| XBDXMDVEZLOGMC-UHFFFAOYSA-N | |
| 1-(chloromethyl)-3-fluorobenzene | |
| 9974 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 456-42-8 | |
| 144.573 | |
| XBDXMDVEZLOGMC-UHFFFAOYSA-N | |
| 9974 | |
| C1=CC(=CC(=C1)F)CCl |
| C7H6ClF | |
| MFCD00000904 | |
| 3-fluorobenzyl chloride, 1-chloromethyl-3-fluorobenzene, m-fluorobenzyl chloride, benzene, 1-chloromethyl-3-fluoro, 3-fluorobenzylchloride, alpha-chloro-3-fluorotoluene, alpha-chloro-m-fluorotoluene, fluorobenzyl-3 chloride, toluene, .alpha.-chloro-m-fluoro, .alpha.-chloro-m-fluorotoluene | |
| 1-(chloromethyl)-3-fluorobenzene |
Safety and Handling
TSCA : Yes