missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl Chloride 95.0+%, TCI America™

Catalog No. F02105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02105G 5 g
F021025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02105G Supplier TCI America Supplier No. F02105G

Chemical Identifiers

CAS 456-42-8
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000904
InChI Key XBDXMDVEZLOGMC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl chloride, 1-chloromethyl-3-fluorobenzene, m-fluorobenzyl chloride, benzene, 1-chloromethyl-3-fluoro, 3-fluorobenzylchloride, alpha-chloro-3-fluorotoluene, alpha-chloro-m-fluorotoluene, fluorobenzyl-3 chloride, toluene, .alpha.-chloro-m-fluoro, .alpha.-chloro-m-fluorotoluene
PubChem CID 9974
IUPAC Name 1-(chloromethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CCl

Specifications

Color Yellow
Boiling Point 180°C
Quantity 5 g
UN Number 2920
Formula Weight 144.57
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzyl Chloride