missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl chloride, 97%

Catalog No. AAA1266506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1266506 5 g
AAA1266514 25 g
AAA1266522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1266506 Supplier Thermo Scientific Chemicals Supplier No. A1266506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 456-42-8 | C7H6ClF | 144.573 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 456-42-8
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000904
InChI Key XBDXMDVEZLOGMC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl chloride, 1-chloromethyl-3-fluorobenzene, m-fluorobenzyl chloride, benzene, 1-chloromethyl-3-fluoro, 3-fluorobenzylchloride, alpha-chloro-3-fluorotoluene, alpha-chloro-m-fluorotoluene, fluorobenzyl-3 chloride, toluene, .alpha.-chloro-m-fluoro, .alpha.-chloro-m-fluorotoluene
PubChem CID 9974
IUPAC Name 1-(chloromethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CCl

Specifications

Density 1.213
Boiling Point 176°C to 177°C
Flash Point 58°C (136°F)
Odor Pungent
Refractive Index 1.512
Quantity 5 g
UN Number UN2920
Beilstein 742265
Formula Weight 144.58
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 207-264-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only