missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl Bromide 97.0+%, TCI America™

Catalog No. F02085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02085G 5 g
F020825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02085G Supplier TCI America Supplier No. F02085G

Chemical Identifiers

CAS 456-41-7
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00000340
InChI Key SCBZBMXPJYMXRC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl bromide, 1-bromomethyl-3-fluorobenzene, 3-fluorobenzylbromide, m-fluorobenzyl bromide, benzene, 1-bromomethyl-3-fluoro, m-fluorobenzylbromide, alpha-bromo-m-fluorotoluene, alpha-bromo-3-fluorotoluene, toluene, .alpha.-bromo-m-fluoro, 1-bromobenzyl-3-fluorobenzene
PubChem CID 68007
IUPAC Name 1-(bromomethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CBr

Specifications

Color Yellow
Boiling Point 200°C
Quantity 5 g
UN Number 3265
Formula Weight 189.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzyl Bromide