missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzyl Alcohol 97.0+%, TCI America™
$77.00 - $308.03
Chemical Identifiers
| CAS | 456-47-3 |
|---|---|
| Molecular Formula | C7H7FO |
| Molecular Weight (g/mol) | 126.13 |
| MDL Number | MFCD00004631 |
| InChI Key | QDHRSLFSDGCJFX-UHFFFAOYSA-N |
| Synonym | 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486 |
| PubChem CID | 68008 |
| IUPAC Name | (3-fluorophenyl)methanol |
| SMILES | C1=CC(=CC(=C1)F)CO |
Chemical Identifiers
| 456-47-3 | |
| 126.13 | |
| QDHRSLFSDGCJFX-UHFFFAOYSA-N | |
| 68008 | |
| C1=CC(=CC(=C1)F)CO |
| C7H7FO | |
| MFCD00004631 | |
| 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486 | |
| (3-fluorophenyl)methanol |
Specifications
| 456-47-3 | |
| 201°C | |
| MFCD00004631 | |
| 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486 | |
| C1=CC(=CC(=C1)F)CO | |
| 126.13 | |
| 126.13 | |
| Liquid |
| Colorless | |
| C7H7FO | |
| 5 g | |
| QDHRSLFSDGCJFX-UHFFFAOYSA-N | |
| (3-fluorophenyl)methanol | |
| 68008 | |
| ≥97.0% (GC) | |
| 3-Fluorobenzyl Alcohol |
Safety and Handling
TSCA : No