missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl alcohol, 98%

Catalog No. AAA1250914
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1250914 25 g
AAA1250909 10 g
AAA1250918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1250914 Supplier Thermo Scientific Chemicals Supplier No. A1250914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 456-47-3 | C7H7FO | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 456-47-3
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004631
InChI Key QDHRSLFSDGCJFX-UHFFFAOYSA-N
Synonym 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
PubChem CID 68008
IUPAC Name (3-fluorophenyl)methanol
SMILES C1=CC(=CC(=C1)F)CO

Specifications

Density 1.166
Boiling Point 201°C
Flash Point 90°C (194°F)
Odor Characteristic
Refractive Index 1.51
Quantity 25 g
Beilstein 2242511
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzyl alcohol
Hazard Category H227-H302+H332
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
EINECSNumber 207-265-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only