missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzotrifluoride 98.0+%, TCI America™
Supplier: TCI America F0390250G
Specifications
| 3-Fluorobenzotrifluoride | |
| Yellow | |
| C7H4F4 | |
| 250 g | |
| 3-fluorobenzotrifluoride, m-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl benzene, alpha,alpha,alpha,3-tetrafluorotoluene, benzene, 1-fluoro-3-trifluoromethyl, trifluoromethyl-3-fluorobenzene, a,a,a,3-tetrafluorotoluene, 1-trifluoromethyl-3-fluorobenzene, 1-fluoro-3-trifluoro-methyl benzene, 3-trifluoromethyl fluorobenzene | |
| FC1=CC=CC(=C1)C(F)(F)F | |
| 164.10 | |
| 164.10 | |
| Liquid |
| 401-80-9 | |
| 101°C | |
| MFCD00000382 | |
| 1993 | |
| GBOWGKOVMBDPJF-UHFFFAOYSA-N | |
| 1-fluoro-3-(trifluoromethyl)benzene | |
| 67867 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 401-80-9 | |
| 164.10 | |
| GBOWGKOVMBDPJF-UHFFFAOYSA-N | |
| 67867 | |
| FC1=CC=CC(=C1)C(F)(F)F |
| C7H4F4 | |
| MFCD00000382 | |
| 3-fluorobenzotrifluoride, m-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl benzene, alpha,alpha,alpha,3-tetrafluorotoluene, benzene, 1-fluoro-3-trifluoromethyl, trifluoromethyl-3-fluorobenzene, a,a,a,3-tetrafluorotoluene, 1-trifluoromethyl-3-fluorobenzene, 1-fluoro-3-trifluoro-methyl benzene, 3-trifluoromethyl fluorobenzene | |
| 1-fluoro-3-(trifluoromethyl)benzene |
Safety and Handling
TSCA : No