missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. F039025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F039025G 25 g
F0390250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F039025G Supplier TCI America Supplier No. F039025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 401-80-9
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.10
MDL Number MFCD00000382
InChI Key GBOWGKOVMBDPJF-UHFFFAOYSA-N
Synonym 3-fluorobenzotrifluoride, m-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl benzene, alpha,alpha,alpha,3-tetrafluorotoluene, benzene, 1-fluoro-3-trifluoromethyl, trifluoromethyl-3-fluorobenzene, a,a,a,3-tetrafluorotoluene, 1-trifluoromethyl-3-fluorobenzene, 1-fluoro-3-trifluoro-methyl benzene, 3-trifluoromethyl fluorobenzene
PubChem CID 67867
IUPAC Name 1-fluoro-3-(trifluoromethyl)benzene
SMILES FC1=CC=CC(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 101°C
Quantity 25 g
UN Number 1993
Formula Weight 164.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzotrifluoride