missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzotrifluoride, 99%

Catalog No. p-7022334
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1505514 25 g
AAA1505522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1505514 Supplier Thermo Scientific Chemicals Supplier No. A1505514
Only null left

CAS: 401-80-9 | C7H4F4 | 164.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401-80-9
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.10
MDL Number MFCD00000382
InChI Key GBOWGKOVMBDPJF-UHFFFAOYSA-N
Synonym 3-fluorobenzotrifluoride, m-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl benzene, alpha,alpha,alpha,3-tetrafluorotoluene, benzene, 1-fluoro-3-trifluoromethyl, trifluoromethyl-3-fluorobenzene, a,a,a,3-tetrafluorotoluene, 1-trifluoromethyl-3-fluorobenzene, 1-fluoro-3-trifluoro-methyl benzene, 3-trifluoromethyl fluorobenzene
PubChem CID 67867
IUPAC Name 1-fluoro-3-(trifluoromethyl)benzene
SMILES FC1=CC=CC(=C1)C(F)(F)F

Specifications

Density 1.303
Boiling Point 101°C to 102°C
Flash Point 7°C (44°F)
Refractive Index 1.401
Quantity 25 g
UN Number UN1993
Beilstein 2045664
Formula Weight 164.1
Percent Purity 99%
Chemical Name or Material 3-Fluorobenzotrifluoride
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-933-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only