missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzonitrile 98.0+%, TCI America™

Catalog No. F02195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02195G 5 g
F021925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02195G Supplier TCI America Supplier No. F02195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 403-54-3
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001797
InChI Key JZTPKAROPNTQQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
PubChem CID 67880
IUPAC Name 3-fluorobenzonitrile
SMILES FC1=CC=CC(=C1)C#N

Specifications

Color Yellow
Boiling Point 183°C
Quantity 5 g
UN Number 3276
Formula Weight 121.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzonitrile