missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzonitrile, 98%

CAS: 403-54-3 | C7H4FN | 121.11 g/mol

$89.67 - $1016.82

Chemical Identifiers

CAS 403-54-3
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001797
InChI Key JZTPKAROPNTQQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
PubChem CID 67880
IUPAC Name 3-fluorobenzonitrile
SMILES FC1=CC=CC(=C1)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2342606
View Documents
Thermo Scientific Chemicals
B2342606
5 g
Each for $89.67
Only null left
 
AAB2342614
View Documents
Thermo Scientific Chemicals
B2342614
25 g
Each for $280.68
Only null left
 
AAB2342622
View Documents
Thermo Scientific Chemicals
B2342622
100 g
Each for $1,016.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

403-54-3
121.11
JZTPKAROPNTQQV-UHFFFAOYSA-N
67880
FC1=CC=CC(=C1)C#N
C7H4FN
MFCD00001797
benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
3-fluorobenzonitrile

Specifications

403-54-3
1.14
67°C (152°F)
C7H4FN
MFCD00001797
UN3276
benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
FC1=CC=CC(=C1)C#N
121.11
121.11
3-Fluorobenzonitrile
-16°C
183°C
Almond-like
1.5055
5 g
1858941
JZTPKAROPNTQQV-UHFFFAOYSA-N
3-fluorobenzonitrile
67880
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 206-963-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only