missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzonitrile, 98%

Catalog No. AAB2342622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2342622 100 g
AAB2342606 5 g
AAB2342614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2342622 Supplier Thermo Scientific Chemicals Supplier No. B2342622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 403-54-3 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 403-54-3
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001797
InChI Key JZTPKAROPNTQQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
PubChem CID 67880
IUPAC Name 3-fluorobenzonitrile
SMILES FC1=CC=CC(=C1)C#N

Specifications

Melting Point -16°C
Density 1.14
Boiling Point 183°C
Flash Point 67°C (152°F)
Odor Almond-like
Refractive Index 1.5055
Quantity 100 g
UN Number UN3276
Beilstein 1858941
Formula Weight 121.11
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzonitrile
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 206-963-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only