missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoic acid, 99%

CAS: 455-38-9 | C7H5FO2 | 140.11 g/mol

$90.56 - $90.56

Chemical Identifiers

CAS 455-38-9
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00002489
InChI Key MXNBDFWNYRNIBH-UHFFFAOYSA-N
Synonym m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192
PubChem CID 9968
ChEBI CHEBI:20021
IUPAC Name 3-fluorobenzoic acid
SMILES OC(=O)C1=CC=CC(F)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119370250
SDS View Documents
Thermo Scientific Chemicals
119370250
25 g Glass bottle
Each for $90.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

455-38-9
140.11
MXNBDFWNYRNIBH-UHFFFAOYSA-N
9968
3-fluorobenzoic acid
C7H5FO2
MFCD00002489
m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192
CHEBI:20021
OC(=O)C1=CC=CC(F)=C1

Specifications

455-38-9
100.0
White to Yellow
99%
C7H5FO2
MFCD00002489
09, 333
Solubility in water: .. Other solubilities: soluble in pyridine
OC(=O)C1=CC=CC(F)=C1
140.11
CHEBI:20021
99%
3-Fluorobenzoic acid, 99%
98.5
121°C to 125°C
Authentic
Glass bottle
FC6H4CO2H
25 g
m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192
MXNBDFWNYRNIBH-UHFFFAOYSA-N
3-fluorobenzoic acid
9968
140.11
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-248-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only