missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoic acid, 99%

Catalog No. AAA1651114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1651114 25 g
AAA1651122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1651114 Supplier Thermo Scientific Chemicals Supplier No. A1651114

CAS: 455-38-9 | C7H5FO2 | 140.11 g/mol

Used in the preparation of regulation of phenolic acid synthesis in Bacillus subtilis. Also used in the bacterial decarboxylation of o-phthalic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of regulation of phenolic acid synthesis in Bacillus subtilis. Also used in the bacterial decarboxylation of o-phthalic acids.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 455-38-9
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00002489
InChI Key MXNBDFWNYRNIBH-UHFFFAOYSA-N
Synonym m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192
PubChem CID 9968
ChEBI CHEBI:20021
IUPAC Name 3-fluorobenzoic acid
SMILES OC(=O)C1=CC=CC(F)=C1

Specifications

Melting Point 122°C to 124°C
Density 1.474
Quantity 25 g
Beilstein 1906920
Solubility Information Very soluble in water.
Formula Weight 140.11
Percent Purity 99%
Chemical Name or Material 3-Fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-248-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only