missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzenesulfonamide 98.0+%, TCI America™

Catalog No. F11415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11415G 5 g
F11411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11415G Supplier TCI America Supplier No. F11415G

Chemical Identifiers

CAS 1524-40-9
Molecular Formula C6H6FNO2S
Molecular Weight (g/mol) 175.177
MDL Number MFCD00042284
InChI Key CRINBBOGNYCAOV-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1-sulfonamide, benzenesulfonamide, 3-fluoro, 3-fluorobenzenesulphonamide, 3-fbsa, 2weo, pubchem11762, acmc-20a5kf, 3-fluoranylbenzenesulfonamide, ksc180k2p, 3-fluorobenzenesulfonamide
PubChem CID 150924
IUPAC Name 3-fluorobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)F

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 175.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorobenzenesulfonamide