missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroanisole 99.0+%, TCI America™

Catalog No. p-7129573
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F025410G 10 g
F025425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F025410G Supplier TCI America Supplier No. F025410G
Only null left

Chemical Identifiers

CAS 456-49-5
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000335
InChI Key MFJNOXOAIFNSBX-UHFFFAOYSA-N
Synonym 3-fluoroanisole, m-fluoroanisole, benzene, 1-fluoro-3-methoxy, 1-fluoro-3-methoxy-benzene, 2-fluoro-4-methoxybenzene, fr co1 wln, 3-fluoroanisol, 3-fluoro-anisole, 3-fluoro anisole, anisole, m-fluoro
PubChem CID 9975
IUPAC Name 1-fluoro-3-methoxybenzene
SMILES COC1=CC=CC(F)=C1

Specifications

Color Yellow
Boiling Point 158°C
Quantity 10 g
UN Number 1993
Formula Weight 126.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluoroanisole