missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroanisole, 98%, Thermo Scientific™

Catalog No. AC171891000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171891000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171891000 Supplier Thermo Scientific Chemicals Supplier No. 171891000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

May darken on storage

Chemical Identifiers

CAS 456-49-5
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000335
InChI Key MFJNOXOAIFNSBX-UHFFFAOYSA-N
Synonym 3-fluoroanisole, m-fluoroanisole, benzene, 1-fluoro-3-methoxy, 1-fluoro-3-methoxy-benzene, 2-fluoro-4-methoxybenzene, fr co1 wln, 3-fluoroanisol, 3-fluoro-anisole, 3-fluoro anisole, anisole, m-fluoro
PubChem CID 9975
IUPAC Name 1-fluoro-3-methoxybenzene
SMILES COC1=CC=CC(F)=C1

Specifications

Melting Point -35.0°C
Density 1.1040g/mL
Boiling Point 158.0°C (743.0 mmHg)
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4876 to 1.4896
Linear Formula FC6H4OCH3
Quantity 100 g
Specific Gravity 1.104
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 3-Fluoroanisole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 207-267-4

RUO – Research Use Only