missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-DL-valine, 94%, Thermo Scientific Chemicals

Catalog No. AAL15334MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15334MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL15334MC Supplier Thermo Scientific Chemicals Supplier No. L15334MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 43163-94-6 | Molecular Formula: C5H10FNO2 | Molecular Weight: 135.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 43163-94-6
Molecular Formula C5H10FNO2
Molecular Weight (g/mol) 135.14
MDL Number MFCD00077549
InChI Key ZFUKCHCGMBNYHH-UHFFFAOYSA-N
Synonym 3-fluoro-dl-valine, 3-fluorovaline, acmc-1cue2
PubChem CID 338887
IUPAC Name 2-amino-3-fluoro-3-methylbutanoic acid
SMILES CC(C)(F)C(N)C(O)=O

Specifications

Melting Point ∽250°C (decomposition)
Beilstein 2350377
Quantity 100 mg
Formula Weight 135.14
Percent Purity 94%
Chemical Name or Material 3-Fluoro-DL-valine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only