accessibility menu, dialog, popup

UserName

CAS RN 43163-94-6

IUPAC Name: 2-amino-3-fluoro-3-methylbutanoic acid
Synonyms: 2-amino-3-fluoro-3-methylbutanoic acid|3-FLUORO-DL-VALINE
Molecular Weight (g/mol): 135.14
Molecular Formula: C5H10FNO2
InChi Key: ZFUKCHCGMBNYHH-UHFFFAOYSA-N
SMILES: CC(C)(F)C(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)