missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-5-(trifluoromethyl)benzaldehyde 98.0+%, TCI America™
$88.56 - $323.43
Chemical Identifiers
| CAS | 188815-30-7 |
|---|---|
| Molecular Formula | C8H4F4O |
| Molecular Weight (g/mol) | 192.113 |
| MDL Number | MFCD00061312 |
| InChI Key | UZZYXZWSOWQPIS-UHFFFAOYSA-N |
| Synonym | 3-fluoro-5-trifluoromethyl benzaldehyde, 3-fluoro-5-formylbenzotrifluoride, alpha,alpha,alpha,5-tetrafluoro-m-tolualdehyde, benzaldehyde, 3-fluoro-5-trifluoromethyl, pubchem4239, acmc-1bzt1, 5-tetrafluoro-m-tolualdehyde, 3-fluoro-5-trifluromethylbenzaldehyde |
| PubChem CID | 605686 |
| IUPAC Name | 3-fluoro-5-(trifluoromethyl)benzaldehyde |
| SMILES | C1=C(C=C(C=C1C(F)(F)F)F)C=O |
Chemical Identifiers
| 188815-30-7 | |
| 192.113 | |
| UZZYXZWSOWQPIS-UHFFFAOYSA-N | |
| 605686 | |
| C1=C(C=C(C=C1C(F)(F)F)F)C=O |
| C8H4F4O | |
| MFCD00061312 | |
| 3-fluoro-5-trifluoromethyl benzaldehyde, 3-fluoro-5-formylbenzotrifluoride, alpha,alpha,alpha,5-tetrafluoro-m-tolualdehyde, benzaldehyde, 3-fluoro-5-trifluoromethyl, pubchem4239, acmc-1bzt1, 5-tetrafluoro-m-tolualdehyde, 3-fluoro-5-trifluromethylbenzaldehyde | |
| 3-fluoro-5-(trifluoromethyl)benzaldehyde |
Specifications
| 188815-30-7 | |
| 175°C | |
| MFCD00061312 | |
| 3-fluoro-5-trifluoromethyl benzaldehyde, 3-fluoro-5-formylbenzotrifluoride, alpha,alpha,alpha,5-tetrafluoro-m-tolualdehyde, benzaldehyde, 3-fluoro-5-trifluoromethyl, pubchem4239, acmc-1bzt1, 5-tetrafluoro-m-tolualdehyde, 3-fluoro-5-trifluromethylbenzaldehyde | |
| C1=C(C=C(C=C1C(F)(F)F)F)C=O | |
| 192.113 | |
| 192.11 | |
| Liquid |
| Yellow | |
| C8H4F4O | |
| 1 g | |
| UZZYXZWSOWQPIS-UHFFFAOYSA-N | |
| 3-fluoro-5-(trifluoromethyl)benzaldehyde | |
| 605686 | |
| ≥98.0% (GC) | |
| 3-Fluoro-5-(trifluoromethyl)benzaldehyde |
Safety and Handling
TSCA : No