missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)benzaldehyde 98.0+%, TCI America™

Catalog No. F07741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07741G 1 g
F07745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07741G Supplier TCI America Supplier No. F07741G

Chemical Identifiers

CAS 188815-30-7
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
MDL Number MFCD00061312
InChI Key UZZYXZWSOWQPIS-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl benzaldehyde, 3-fluoro-5-formylbenzotrifluoride, alpha,alpha,alpha,5-tetrafluoro-m-tolualdehyde, benzaldehyde, 3-fluoro-5-trifluoromethyl, pubchem4239, acmc-1bzt1, 5-tetrafluoro-m-tolualdehyde, 3-fluoro-5-trifluromethylbenzaldehyde
PubChem CID 605686
IUPAC Name 3-fluoro-5-(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C=C1C(F)(F)F)F)C=O

Specifications

Color Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 192.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzaldehyde