missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzonitrile 99.0+%, TCI America™

Catalog No. F10191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10191G 1 g
F10195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10191G Supplier TCI America Supplier No. F10191G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 231953-38-1
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.113
MDL Number MFCD00236280
InChI Key CFPNGJCBTZAVDN-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethyl benzonitrile, 4-cyano-2-fluorobenzotrifluoride, benzonitrile, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl benzenecarbonitrile, alpha,alpha,alpha,3-tetrafluoro-p-tolunitrile, pubchem4800, acmc-1cq1l, intermediates-zcf02625, ksc495o7j, 3-fluoro-4-trifluoromethyl-benzonitrile
PubChem CID 2737550
IUPAC Name 3-fluoro-4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C#N)F)C(F)(F)F

Specifications

Melting Point 48°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 189.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzonitrile