missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. F07581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07581G 1 g
F07585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07581G Supplier TCI America Supplier No. F07581G

Chemical Identifiers

CAS 115754-21-7
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD00236279
InChI Key HRIHSNPFVGMAKX-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethyl benzoic acid, benzoic acid, 3-fluoro-4-trifluoromethyl, alpha,alpha,alpha,3-tetrafluoro-p-toluic acid, pubchem4970, acmc-1c70e, ksc179k4t, 3-fluoro-4 trifluoromethyl benzoic acid, 3-fluoro-4-trifluoromethyl-benzoic acid, 3-fluoro-4-trifluoromethyl benzoicacid, 3-fluoro-4-trifluoromethyl benzoic aicd
PubChem CID 688293
IUPAC Name 3-fluoro-4-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)C(F)(F)F

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoic Acid