missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)aniline, 97%

Catalog No. p-7027564
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2441103 1 g
AAB2441106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2441103 Supplier Thermo Scientific Chemicals Supplier No. B2441103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69411-68-3 | C7H5F4N | 179.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69411-68-3
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00190120
InChI Key CRRVZRDISHOQQL-UHFFFAOYSA-N
Synonym 4-amino-2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl aniline, 2-fluoro-4-amino benzotrifluoride, benzenamine, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl-phenylamine, 3-fluoro-4-trifluoromethyl phenylamine, pubchem1631, 4-amino,2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl-aniline
PubChem CID 2735914
IUPAC Name 3-fluoro-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C(F)=C1)C(F)(F)F

Specifications

Melting Point 54°C to 58°C
Quantity 1 g
UN Number UN2811
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)aniline
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only