missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4'-propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F09421G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09421G Supplier TCI America Supplier No. F09421G

Specifications

Chemical Name or Material 3-Fluoro-4′-propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride)
CAS 909709-42-8
Color White-Yellow
Molecular Formula C15H16BFO2
Quantity 1 g
MDL Number MFCD10687198
Synonym 3-fluoro-4'-propyl-1,1'-biphenyl-4-yl boronic acid, 4'-propyl-3-fluorobiphenyl-4-boronic acid, 4-propyl-3-fluorobiphenyl-4-boronic acid, 3-fluoro-4'-propyl-biphenylboronic acid, 3-fluoro-4'-propyl-4-biphenylboronic acid, 4'-propyl-3-fluoro-4-biphenyl boronic acid, 2-fluoro-4-4-propylphenyl phenylboronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl
InChI Key XZGCBSYWWZAXRC-UHFFFAOYSA-N
SMILES CCCC1=CC=C(C=C1)C1=CC(F)=C(C=C1)B(O)O
IUPAC Name {3-fluoro-4'-propyl-[1,1'-biphenyl]-4-yl}boronic acid
Molecular Weight (g/mol) 258.10
PubChem CID 53402576
Formula Weight 258.10
Physical Form Crystalline Powder
Show More Show Less