missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrotoluene 98.0+%, TCI America™

Catalog No. p-7129758
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05675G 5 g
F056725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05675G Supplier TCI America Supplier No. F05675G
Only null left

Chemical Identifiers

CAS 446-34-4
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007053
InChI Key WZMOWQCNPFDWPA-UHFFFAOYSA-N
Synonym 3-fluoro-4-nitrotoluene, benzene, 2-fluoro-4-methyl-1-nitro, 2-fluoro-4-methylnitrobenzene, 2-fluoro-4-methyl-1-nitro-benzene, 3-fluoro-4-nitrobenzene, 2-fluor-4-methyl-1-nitrobenzol, pubchem1595, 3-fluoro4-nitrotoluene, acmc-1alaq, 3-fluoro-4-nitro toluene
PubChem CID 67966
IUPAC Name 2-fluoro-4-methyl-1-nitrobenzene
SMILES CC1=CC=C(C(F)=C1)[N+]([O-])=O

Specifications

Melting Point 54°C
Color Yellow
Boiling Point 98°C
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-nitrotoluene