missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrophenol 98.0+%, TCI America™

Catalog No. p-7129628
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
F03485G 5 g
F034825G 25 g
2 options

Catalog No. F03485G

Supplier: TCI America F03485G

Only null left

Chemical Identifiers

CAS 394-41-2
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD00041251
InChI Key CSSGKHVRDGATJL-UHFFFAOYSA-N
Synonym phenol, 3-fluoro-4-nitro, 3-fluoro-4-nitro-phenol, 2-fluoro-4-hydroxynitrobenzene, pubchem2101, 3-fluoro4-nitrophenol, 4-nitro-3-fluorophenol, 5-fluoro-4-nitrophenol, acmc-1afpw, 3-fluoro-4-nitro phenol, ksc490k8p
PubChem CID 520948
IUPAC Name 3-fluoro-4-nitrophenol
SMILES C1=CC(=C(C=C1O)F)[N+](=O)[O-]

Specifications

Melting Point 92°C
Quantity 5 g
Formula Weight 157.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-nitrophenol