missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrophenol, 99%, Thermo Scientific Chemicals

Catalog No. p-7021844
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1435006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1435006 Supplier Thermo Scientific Chemicals Supplier No. A1435006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 394-41-2 | Molecular Formula: C6H4FNO3 | Molecular Weight: 157.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 394-41-2
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD00041251
InChI Key CSSGKHVRDGATJL-UHFFFAOYSA-N
Synonym phenol, 3-fluoro-4-nitro, 3-fluoro-4-nitro-phenol, 2-fluoro-4-hydroxynitrobenzene, pubchem2101, 3-fluoro4-nitrophenol, 4-nitro-3-fluorophenol, 5-fluoro-4-nitrophenol, acmc-1afpw, 3-fluoro-4-nitro phenol, ksc490k8p
PubChem CID 520948
IUPAC Name 3-fluoro-4-nitrophenol
SMILES C1=CC(=C(C=C1O)F)[N+](=O)[O-]

Specifications

Melting Point 91°C to 93°C
Quantity 5 g
Beilstein 2048033
Formula Weight 157.1
Percent Purity 99%
Chemical Name or Material 3-Fluoro-4-nitrophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 206-895-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only