missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-morpholinoaniline 98.0+%, TCI America™
Supplier: TCI America F084925G
Specifications
| 3-Fluoro-4-morpholinoaniline | |
| 123°C | |
| MFCD02571270 | |
| 4-(2-Fluoro-4-aminophenyl)morpholine | |
| NC1=CC(F)=C(C=C1)N1CCOCC1 | |
| 196.23 | |
| 196.23 | |
| Crystalline Powder |
| 93246-53-8 | |
| C10H13FN2O | |
| 25 g | |
| FQGIBHQUVCGEAC-UHFFFAOYSA-N | |
| 3-fluoro-4-(morpholin-4-yl)aniline | |
| 1485330 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 93246-53-8 | |
| 196.23 | |
| FQGIBHQUVCGEAC-UHFFFAOYSA-N | |
| 1485330 | |
| NC1=CC(F)=C(C=C1)N1CCOCC1 |
| C10H13FN2O | |
| MFCD02571270 | |
| 4-(2-Fluoro-4-aminophenyl)morpholine | |
| 3-fluoro-4-(morpholin-4-yl)aniline |
Safety and Handling
TSCA : No