missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylaniline 98.0+%, TCI America™

Catalog No. F04405G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04405G Supplier TCI America Supplier No. F04405G
Only null left

Specifications

CAS 452-77-7
Molecular Formula C7H8FN
Quantity 5 g
MDL Number MFCD00007762
UN Number 2811
Synonym 4-amino-2-fluorotoluene, 3-fluoro-p-toluidine, 2-fluoro-4-aminotoluene, 2-fluoro-4-amino-toluene, benzenamine, 3-fluoro-4-methyl, p-toluidine, 3-fluoro, 3-fluoro-4-methylphenylamine, 3-fluoro-4-methyl aniline, 3-fluoro-4-methyl-aniline, benzenamine, 3-fluoro-4-methyl-9ci
InChI Key MGRHBBRSAFPBIN-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)N)F
IUPAC Name 3-fluoro-4-methylaniline
Molecular Weight (g/mol) 125.146
PubChem CID 9957
Formula Weight 125.15
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Fluoro-4-methylaniline
Show More Show Less
RTECSNumber XU6825000
TSCA No