missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$61.51 - $61.51
Chemical Identifiers
| CAS | 149507-26-6 |
|---|---|
| Molecular Formula | C7H8BFO3 |
| Molecular Weight (g/mol) | 169.946 |
| MDL Number | MFCD00807404 |
| InChI Key | IILGLPAJXQMKGQ-UHFFFAOYSA-N |
| PubChem CID | 2782194 |
| IUPAC Name | (3-fluoro-4-methoxyphenyl)boronic acid |
| SMILES | B(C1=CC(=C(C=C1)OC)F)(O)O |
Chemical Identifiers
| 149507-26-6 | |
| 169.946 | |
| IILGLPAJXQMKGQ-UHFFFAOYSA-N | |
| (3-fluoro-4-methoxyphenyl)boronic acid |
| C7H8BFO3 | |
| MFCD00807404 | |
| 2782194 | |
| B(C1=CC(=C(C=C1)OC)F)(O)O |
Specifications
| 149507-26-6 | |
| C7H8BFO3 | |
| 1 g | |
| IILGLPAJXQMKGQ-UHFFFAOYSA-N | |
| (3-fluoro-4-methoxyphenyl)boronic acid | |
| 2782194 | |
| Crystalline Powder |
| 198°C | |
| MFCD00807404 | |
| 3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid, boronic acid, 3-fluoro-4-methoxyphenyl, 3-fluoro-4-methoxybenzeneboronicacid, pubchem1850, 3-fluoro-4-methoxy-benzeneboronic acid | |
| B(C1=CC(=C(C=C1)OC)F)(O)O | |
| 169.946 | |
| 169.95 | |
| 3-Fluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No