accessibility menu, dialog, popup

UserName

H3COFBOHHO

CAS RN 149507-26-6

IUPAC Name: (3-fluoro-4-methoxyphenyl)boronic acid
Synonyms: 3-fluoro-4-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: IILGLPAJXQMKGQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1F)B(O)O
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)