missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F08761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08761G Supplier TCI America Supplier No. F08761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149507-26-6
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD00807404
InChI Key IILGLPAJXQMKGQ-UHFFFAOYSA-N
PubChem CID 2782194
IUPAC Name (3-fluoro-4-methoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OC)F)(O)O

Specifications

Melting Point 198°C
Quantity 1 g
Synonym 3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid, boronic acid, 3-fluoro-4-methoxyphenyl, 3-fluoro-4-methoxybenzeneboronicacid, pubchem1850, 3-fluoro-4-methoxy-benzeneboronic acid
Formula Weight 169.95
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride)