missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzyl Chloride 98.0+%, TCI America™

Catalog No. p-7130000
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09665G 5 g
F096625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09665G Supplier TCI America Supplier No. F09665G

Chemical Identifiers

CAS 351-52-0
Molecular Formula C8H8ClFO
Molecular Weight (g/mol) 174.60
MDL Number MFCD04973773
InChI Key DDAXEANMRGIVDY-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzyl chloride, 4-chloromethyl-2-fluoro-1-methoxybenzene, 3-fluoro-4-methoxybenzylchloride, benzene,4-chloromethyl-2-fluoro-1-methoxy, benzene, 4-chloromethyl-2-fluoro-1-methoxy, 4-chloromethyl-2-fluorophenyl methyl ether, 4-chloromethyl-2-fluoro-1-methoxy-benzene, 3-fluor-4-methoxy-benzylchlorid, 4-chloromethyl-2-fluoroanisole, 3-fluoro-4-methoxy benzyl chloride
PubChem CID 2060903
IUPAC Name 4-(chloromethyl)-2-fluoro-1-methoxybenzene
SMILES COC1=CC=C(CCl)C=C1F

Specifications

Color White-Yellow
Boiling Point 62°C
Quantity 5 g
UN Number 3261
Formula Weight 174.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Fluoro-4-methoxybenzyl Chloride