missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzyl chloride, 98%

Numéro de catalogue. p-7030872
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH3175603 1 g
AAH3175606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH3175603 Fournisseur Thermo Scientific Chemicals Code fournisseur H3175603
Il en reste null

CAS: 351-52-0 | C8H8ClFO | 174.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 351-52-0
Molecular Formula C8H8ClFO
Molecular Weight (g/mol) 174.60
MDL Number MFCD04973773
InChI Key DDAXEANMRGIVDY-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzyl chloride, 4-chloromethyl-2-fluoro-1-methoxybenzene, 3-fluoro-4-methoxybenzylchloride, benzene,4-chloromethyl-2-fluoro-1-methoxy, benzene, 4-chloromethyl-2-fluoro-1-methoxy, 4-chloromethyl-2-fluorophenyl methyl ether, 4-chloromethyl-2-fluoro-1-methoxy-benzene, 3-fluor-4-methoxy-benzylchlorid, 4-chloromethyl-2-fluoroanisole, 3-fluoro-4-methoxy benzyl chloride
PubChem CID 2060903
IUPAC Name 4-(chloromethyl)-2-fluoro-1-methoxybenzene
SMILES COC1=CC=C(CCl)C=C1F

Spécifications

Melting Point 35°C to 39°C
Quantity 1 g
UN Number UN3261
Formula Weight 174.6
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only