missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzyl chloride, 98%

Catalog No. AAH3175606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3175606 5 g
AAH3175603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3175606 Supplier Thermo Scientific Chemicals Supplier No. H3175606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 351-52-0 | C8H8ClFO | 174.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 351-52-0
Molecular Formula C8H8ClFO
Molecular Weight (g/mol) 174.60
MDL Number MFCD04973773
InChI Key DDAXEANMRGIVDY-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzyl chloride, 4-chloromethyl-2-fluoro-1-methoxybenzene, 3-fluoro-4-methoxybenzylchloride, benzene,4-chloromethyl-2-fluoro-1-methoxy, benzene, 4-chloromethyl-2-fluoro-1-methoxy, 4-chloromethyl-2-fluorophenyl methyl ether, 4-chloromethyl-2-fluoro-1-methoxy-benzene, 3-fluor-4-methoxy-benzylchlorid, 4-chloromethyl-2-fluoroanisole, 3-fluoro-4-methoxy benzyl chloride
PubChem CID 2060903
IUPAC Name 4-(chloromethyl)-2-fluoro-1-methoxybenzene
SMILES COC1=CC=C(CCl)C=C1F

Specifications

Melting Point 35°C to 39°C
Quantity 5 g
UN Number UN3261
Formula Weight 174.6
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only