missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-methoxybenzoic Acid 98.0+%, TCI America™
$127.07 - $417.77
Chemical Identifiers
| CAS | 403-20-3 |
|---|---|
| Molecular Formula | C8H6FO3 |
| Molecular Weight (g/mol) | 169.13 |
| MDL Number | MFCD00060675 |
| InChI Key | HYNNNQDQEORWEU-UHFFFAOYSA-M |
| Synonym | 3-fluoro-4-methoxy-benzoic acid, 3-fluoro-p-anisic acid, 3-fluoro-4-methoxy benzoic acid, 3-fluoro-4-methoxybenzoicacid, pubchem1393, opera_id_903, acmc-209jd1, ksc494s2f, 3-fluoro-4-methoxy benzoicacid, rarechem al bo 0870 |
| PubChem CID | 2733401 |
| IUPAC Name | 3-fluoro-4-methoxybenzoate |
| SMILES | COC1=CC=C(C=C1F)C([O-])=O |
Chemical Identifiers
| 403-20-3 | |
| 169.13 | |
| HYNNNQDQEORWEU-UHFFFAOYSA-M | |
| 2733401 | |
| COC1=CC=C(C=C1F)C([O-])=O |
| C8H6FO3 | |
| MFCD00060675 | |
| 3-fluoro-4-methoxy-benzoic acid, 3-fluoro-p-anisic acid, 3-fluoro-4-methoxy benzoic acid, 3-fluoro-4-methoxybenzoicacid, pubchem1393, opera_id_903, acmc-209jd1, ksc494s2f, 3-fluoro-4-methoxy benzoicacid, rarechem al bo 0870 | |
| 3-fluoro-4-methoxybenzoate |
Specifications
| 403-20-3 | |
| White-Yellow | |
| MFCD00060675 | |
| 3-fluoro-4-methoxy-benzoic acid, 3-fluoro-p-anisic acid, 3-fluoro-4-methoxy benzoic acid, 3-fluoro-4-methoxybenzoicacid, pubchem1393, opera_id_903, acmc-209jd1, ksc494s2f, 3-fluoro-4-methoxy benzoicacid, rarechem al bo 0870 | |
| COC1=CC=C(C=C1F)C([O-])=O | |
| 169.13 | |
| 170.14 | |
| Crystalline Powder |
| 212°C | |
| C8H6FO3 | |
| 1 g | |
| HYNNNQDQEORWEU-UHFFFAOYSA-M | |
| 3-fluoro-4-methoxybenzoate | |
| 2733401 | |
| ≥98.0% (GC,T) | |
| 3-Fluoro-4-methoxybenzoic Acid |
Safety and Handling
TSCA : No