missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzeneboronic acid, 98+%

Catalog No. AAL1981803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1981803 1 g
AAL1981806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1981803 Supplier Thermo Scientific Chemicals Supplier No. L1981803

CAS: 149507-26-6 | C7H8BFO3 | 169.946 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149507-26-6
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD00807404
InChI Key IILGLPAJXQMKGQ-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid, boronic acid, 3-fluoro-4-methoxyphenyl, 3-fluoro-4-methoxybenzeneboronicacid, pubchem1850, 3-fluoro-4-methoxy-benzeneboronic acid
PubChem CID 2782194
IUPAC Name (3-fluoro-4-methoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OC)F)(O)O

Specifications

Melting Point 237°C to 239°C
Quantity 1 g
Beilstein 7370120
Formula Weight 169.95
Percent Purity ≥98%
Chemical Name or Material 3-Fluoro-4-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only