missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxyaniline 98.0+%, TCI America™

Catalog No. p-7129808
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06485G 5 g
F064825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06485G Supplier TCI America Supplier No. F06485G

Chemical Identifiers

CAS 366-99-4
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.145
MDL Number MFCD00075040
InChI Key LJWAPDSCYTZUJU-UHFFFAOYSA-N
Synonym 3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine
PubChem CID 581110
IUPAC Name 3-fluoro-4-methoxyaniline
SMILES COC1=C(C=C(C=C1)N)F

Specifications

Melting Point 82°C
Boiling Point 135°C
Quantity 5 g
Formula Weight 141.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-methoxyaniline