missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxyaniline, 98+%

Catalog No. AAA1860606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1860606 5 g
AAA1860614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1860606 Supplier Thermo Scientific Chemicals Supplier No. A1860606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 366-99-4 | C7H8FNO | 141.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 366-99-4
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.145
MDL Number MFCD00075040
InChI Key LJWAPDSCYTZUJU-UHFFFAOYSA-N
Synonym 3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine
PubChem CID 581110
IUPAC Name 3-fluoro-4-methoxyaniline
SMILES COC1=C(C=C(C=C1)N)F

Specifications

Melting Point 80°C to 84°C
Boiling Point 135°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2811
Beilstein 2717083
Formula Weight 141.15
Percent Purity ≥98%
Chemical Name or Material 3-Fluoro-4-methoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only