missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-iodoaniline, 98%

Catalog No. p-7053766
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6142703 1 g
AAH6142706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6142703 Supplier Thermo Scientific Chemicals Supplier No. H6142703

CAS: 656-66-6 | C6H5FIN | 237.016 g/mol

3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water (228.3 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

Chemical Identifiers

CAS 656-66-6
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD07774187
InChI Key KUVVJHBHRIXJKI-UHFFFAOYSA-N
Synonym 3-fluoro-4-iodophenylamine, benzenamine, 3-fluoro-4-iodo, 3-fluoro-4-iodo-aniline, zlchem 541, pubchem1534, 4-iodo 3-fluoro aniline, acmc-1b9hq, 3-fluoro-4-iodo-phenylamine, buttpark 120\07-56
PubChem CID 15475546
IUPAC Name 3-fluoro-4-iodoaniline
SMILES C1=CC(=C(C=C1N)F)I

Specifications

Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Solubility Information Slightly soluble in water (228.3mg/L at 25°C).
Formula Weight 237.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Fluoro-4-iodoaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only